missing translation for 'onlineSavingsMsg'
Learn More

AP3M2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-56624

 View more versions of this product

Product Code. 18218352

  • £362.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AP3M2 Polyclonal specifically detects AP3M2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AP3M2
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
Adapter-related protein complex 3 mu-2 subunit, adaptor-related protein complex 3, mu 2 subunit, AP-3 complex subunit mu-2, AP47B, CLA20, Clathrin assembly protein assembly protein complex 1 medium chain homolog 2, Clathrin coat assembly protein AP47 homolog 2, Clathrin coat-associated protein AP47 homolog 2, clathrin-associated protein AP47 homolog 2, Golgi adaptor AP-1 47 kDa protein homolog 2, HA1 47 kDa subunit homolog 2, HA1 47kDA subunit homolog 2, mu3B-adaptin, P47B
Rabbit
47 kDa
100 μL
Primary
This product is specific to Subunit or Isoform: mu-2.
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
IgG
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
P53677
AP3M2
Synthetic peptides corresponding to AP3M2(adaptor-related protein complex 3, mu 2 subunit) The peptide sequence was selected from the middle region of AP3M2 (NP_006794). Peptide sequence VVNTITGSTNVGDQLPTGQLSVVPWRRTGVKYTNNEAYFDVIEEIDAIID.
Affinity purified
RUO
10947
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.