missing translation for 'onlineSavingsMsg'
Learn More

AP3M1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-76589-0.025mg

 View more versions of this product

Product Code. 18593712

  • £150.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AP3M1 Polyclonal specifically detects AP3M1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AP3M1
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
adaptor-related protein complex 3, mu 1 subunit, AP-3 adapter complex mu3A subunit, MGC22164, mu-3A subunit, mu3A-adaptin
Rabbit
47s kDa
0.025 mg
Primary
AP3M1 antibody may cross-react with AP3M2.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Western Blot
1 mg/mL
Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL
Q9Y2T2
AP3M1
Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human AP3M1. The immunogen is located within amino acids 340 - 390 of AP3M1. Amino Acid Squence: VNLQSGAPKPEENPSLN
Affinity Purified
RUO
26985
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only