missing translation for 'onlineSavingsMsg'
Learn More

AP3B2 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18425787 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425787 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425787 Supplier Novus Biologicals Supplier No. NBP176590AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AP3B2 Polyclonal antibody specifically detects AP3B2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP3B2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias Adapter-related protein complex 3 subunit beta-2, Adaptor protein complex AP-3 subunit beta-2, adaptor-related protein complex 3, beta 2 subunit, beta-3B-adaptin, Clathrin assembly protein complex 3 beta-2 large chain, DKFZp686D17136, NAPTBAP-3 complex subunit beta-2, Neuronal adaptin-like protein, beta-subunit, Neuron-specific vesicle coat protein beta-NAP
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human AP3B2. The immunogen is located within the first 50 amino acids of AP3B2. Amino Acid Squence: IFSSDYKRHDDLKEMLDT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8120
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.