missing translation for 'onlineSavingsMsg'
Learn More

AP3B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13384-1-AP-20UL

 View more versions of this product

Product Code. 16881564

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. The encoded protein is part of the heterotetrameric AP-3 protein complex which interacts with the scaffolding protein clathrin. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 2.
Specifications

Specifications

AP3B1
Polyclonal
Unconjugated
AP3B1
ADTB3, ADTB3A, AP 3 complex subunit beta 1, AP3B1, Beta 3A adaptin, HPS, HPS2, PE
Rabbit
Antigen Affinity Chromatography
RUO
11774, 309969, 8546
-20°C
Liquid
Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00203, Q9Z1T1
Ap3b1
AP3B1 Fusion Protein Ag4225
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.