missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP3B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13384-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. The encoded protein is part of the heterotetrameric AP-3 protein complex which interacts with the scaffolding protein clathrin. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 2.Specifications
| AP3B1 | |
| Polyclonal | |
| Unconjugated | |
| AP3B1 | |
| ADTB3, ADTB3A, AP 3 complex subunit beta 1, AP3B1, Beta 3A adaptin, HPS, HPS2, PE | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11774, 309969, 8546 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00203, Q9Z1T1 | |
| Ap3b1 | |
| AP3B1 Fusion Protein Ag4225 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction