missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AP3B1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13464187
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13464187 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13464187 Supplier Invitrogen™ Supplier No. PA529858

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, HepG2. Predicted reactivity: Mouse (98%), Xenopus laevis (96%), Dog (100%), Pig (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

AP3B1 is a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Structurally, AP3B1 is part of the heterotetrameric AP-3 protein complex which interacts with the scaffolding protein clathrin. Mutations in AP3B1 are associated with Hermansky-Pudlak syndrome type 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP3B1
Applications Western Blot
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Ap3b1
Gene Accession No. O00203, Q9Z1T1
Gene Alias adapter-related protein complex 3 subunit beta-1; adaptor protein complex AP-3 beta-1 subunit; adaptor protein complex AP-3 subunit beta-1; adaptor related protein complex 3 beta 1 subunit; adaptor related protein complex 3 subunit beta 1; adaptor related protein complex 3, beta 1 subunit; adaptor-related protein complex 3 subunit beta-1; adaptor-related protein complex 3, beta 1 subunit; adaptor-related protein complex AP3 beta 1 subunit; adaptor-related protein complex AP-3, beta 1 subunit; ADTB3; ADTB3A; ap3; AP-3; AP-3 complex beta-3A subunit; AP-3 complex subunit beta-1; AP-3 complex subunit beta-1-like protein; AP3B1; AU015684; beta3A; Beta-3A-adaptin; beta3A-adaptin; C78395; Clathrin assembly protein complex 3 beta-1 large chain; HPS; Hps2; PE; pearl; recombination induced mutation 2; rim2; Unknown (protein for MGC:128992)
Gene Symbols Ap3b1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human HPS2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11774, 8546
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.