missing translation for 'onlineSavingsMsg'
Learn More

AP2S1 Antibody (3E4), Novus Biologicals™

Product Code. 18341238 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341238 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341238 Supplier Novus Biologicals Supplier No. H00001175M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AP2S1 Monoclonal antibody specifically detects AP2S1 in Human samples. It is validated for Western Blot, ELISA, ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP2S1
Applications Western Blot, ELISA, Sandwich ELISA, KnockDown
Classification Monoclonal
Clone 3E4
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH06337
Gene Alias Adapter-related protein complex 2 sigma subunit, Adaptor protein complex AP-2 subunit sigma, adaptor-related protein complex 2, sigma 1 subunit, AP17HA2 17 kDa subunit, AP-2 complex subunit sigma, CLAPS2AP17-DELTA, Clathrin assembly protein 2 small chain, Clathrin coat assembly protein AP17, Clathrin coat-associated protein AP17, clathrin-associated/assembly/adaptor protein, small 2 (17kD), Plasma membrane adaptor AP-2 17 kDa protein, Sigma2-adaptin
Host Species Mouse
Immunogen AP2S1 (AAH06337, 1 a.a. ~ 142 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MIRFILIQNRAGKTRLAKWYMQFDDDEKQKLIEEVHAVVTVRDAKHTNFVEFRNFKIIYRRYAGLYFCICVDVNDNNLAYLEAIHNFVEVLNEYFHNVCELDLVFNFYKVYTVVDEMFLAGEIRETSQTKVLKQLLMLQSLE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1175
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.