missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565347
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: SGLLVDVFSDS ASVVAPLAPG SEDNFARFVC K Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
Adaptins are components of the adaptor complexes which link clathrin to receptors in coated vesicles. Clathrin-associated protein complexes are believed to interact with the cytoplasmic tails of membrane proteins, leading to their selection and concentration. Alpha adaptin is a subunit of the plasma membrane adaptor. It binds polyphosphoinositides.Specifications
AP2A2 | |
Polyclonal | |
Unconjugated | |
AP2A2 | |
AP2A2; ADTAB; CLAPA2; HIP9; HYPJ; KIAA0899; AP-2 complex subunit alpha-2; 100 kDa coated vesicle protein C; Adapter-related protein complex 2 alpha-2 subunit; Adaptor protein complex AP-2 subunit alpha-2; Alpha-adaptin C; Alpha2-adaptin; Clathrin assembly | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
161 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94973 | |
AP2A2 | |
Recombit Human AP2A2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only