missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AP2A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576733
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Adaptins are components of the adaptor complexes which link clathrin to receptors in coated vesicles. Clathrin-associated protein complexes are believed to interact with the cytoplasmic tails of membrane proteins, leading to their selection and concentration. Alpha adaptin is a subunit of the plasma membrane adaptor. It binds polyphosphoinositides.
Specifications
| AP2A2 | |
| Polyclonal | |
| Unconjugated | |
| AP2A2 | |
| 100 kDa coated vesicle protein C; 2410074K14Rik; adapter-related protein complex 2 alpha-2 subunit; adapter-related protein complex 2 subunit alpha-2; adaptin alpha A; Adaptin alpha B; adaptin, alpha B; adaptor protein complex AP-2 subunit alpha-2; adaptor protein complex AP-2, alpha 2 subunit; adaptor related protein complex 2 alpha 2 subunit; adaptor related protein complex 2, alpha 2 subunit; adaptor-related protein complex 2 subunit alpha-2; adaptor-related protein complex 2, alpha 2 subunit; adaptor-related protein complex AP-2, alpha 2 subunit; ADTAA; ADTAB; AF006990; Alpha adaptin A; Alpha adaptin C; Alpha1 adaptin; Alpha2 adaptin; alpha2-adaptin; alpha-adaptin C; alpha-adaptin C; Huntingtin interacting protein J; alpha-C adaptin; AP2 alpha; AP-2 complex subunit alpha-2; AP2A1; Ap2a2; AW146353; C78001; CLAPA1; CLAPA2; Clathrin assembly; Clathrin assembly protein complex 2 alpha-C large chain; clathrin-associated/assembly/adaptor protein, large, alpha 2; HIP 9; HIP9; HIP-9; Huntingtin yeast partner J; huntingtin-interacting protein 9; Huntingtin-interacting protein J; HYPJ; KIAA0899; L25; Plasma membrane adaptor HA2/AP2 adaptin alpha C subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11772, 161 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94973, P17427 | |
| AP2A2 | |
| Recombinant full length Human AP2A2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction