missing translation for 'onlineSavingsMsg'
Learn More

AP2A2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-12692-50ul

 View more versions of this product

Product Code. 18691899

  • £161.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AP2A2 Polyclonal antibody specifically detects AP2A2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AP2A2
Polyclonal
Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of uLysate, Immunohistochemistry-Paraffin 1:50-1:200
100 kDa coated vesicle protein C, Adapter-related protein complex 2 alpha-2 subunit, adaptin, alpha B, Adaptor protein complex AP-2 subunit alpha-2, adaptor-related protein complex 2, alpha 2 subunit, alpha2-adaptin, Alpha-adaptin C, alpha-adaptin C; Huntingtin interacting protein J, AP-2 complex subunit alpha-2, Clathrin assembly protein complex 2 alpha-C large chain, clathrin-associated/assembly/adaptor protein, large, alpha 2, DKFZP564D1864, HIP9ADTAB, Huntingtin yeast partner J, Huntingtin-interacting protein 9, Huntingtin-interacting protein J, HYPJCLAPA2, KIAA0899HIP-9, Plasma membrane adaptor HA2/AP2 adaptin alpha C subunit
Produced in rabbits immunized with E. coli-derived Human AP2A2 fragment
50 μg
Neurodegeneration, Neuroscience
161
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
PBS, pH7.0
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.