missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | AP2A2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611156
|
Novus Biologicals
NBP2-99239-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18658325
|
Novus Biologicals
NBP2-99239-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AP2A2 Polyclonal antibody specifically detects AP2A2 in Human samples. It is validated for Western BlotSpecifications
| AP2A2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neurodegeneration, Neuroscience | |
| 0.2 um filtered solution in PBS | |
| 161 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 100 kDa coated vesicle protein C, Adapter-related protein complex 2 alpha-2 subunit, adaptin, alpha B, Adaptor protein complex AP-2 subunit alpha-2, adaptor-related protein complex 2, alpha 2 subunit, alpha2-adaptin, Alpha-adaptin C, alpha-adaptin C; Huntingtin interacting protein J, AP-2 complex subunit alpha-2, Clathrin assembly protein complex 2 alpha-C large chain, clathrin-associated/assembly/adaptor protein, large, alpha 2, DKFZP564D1864, HIP9ADTAB, Huntingtin yeast partner J, Huntingtin-interacting protein 9, Huntingtin-interacting protein J, HYPJCLAPA2, KIAA0899HIP-9, Plasma membrane adaptor HA2/AP2 adaptin alpha C subunit | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human AP2A2. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title