missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2A2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92716-0.02ml
This item is not returnable.
View return policy
Description
AP2A2 Polyclonal antibody specifically detects AP2A2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| AP2A2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| 100 kDa coated vesicle protein C, Adapter-related protein complex 2 alpha-2 subunit, adaptin, alpha B, Adaptor protein complex AP-2 subunit alpha-2, adaptor-related protein complex 2, alpha 2 subunit, alpha2-adaptin, Alpha-adaptin C, alpha-adaptin C; Huntingtin interacting protein J, AP-2 complex subunit alpha-2, Clathrin assembly protein complex 2 alpha-C large chain, clathrin-associated/assembly/adaptor protein, large, alpha 2, DKFZP564D1864, HIP9ADTAB, Huntingtin yeast partner J, Huntingtin-interacting protein 9, Huntingtin-interacting protein J, HYPJCLAPA2, KIAA0899HIP-9, Plasma membrane adaptor HA2/AP2 adaptin alpha C subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 590-939 of human Alpha2-adaptin/AP2A2 (NP_036437.1). STDILATVLEEMPPFPERESSILAKLKKKKGPSTVTDLEDTKRDRSVDVNGGPEPAPASTSAVSTPSPSADLLGLGAAPPAPAGPPPSSGGSGLLVDVFSDSASVVAPLAPGSEDNFARFVCKNNGVLFENQLLQIGLKSEFRQNLGRMFIFYGNKTSTQFLNFTPTLICSDDLQPNLNLQTKPVDPTVEGGAQVQQVVNIECVSDFTEAPVLNIQFRYGGTFQNVSVQLPITLNKFFQPTEMASQDFFQRWKQLSNPQQEVQNIFKAKHPMDTEVTKAKIIGFGSALLEEVDPNPANFVGAGIIHTKTTQIGCLLRLEPNLQAQMYRLTLRTSKEAVSQRLCELLSAQF | |
| 0.02 mL | |
| Neurodegeneration, Neuroscience | |
| 161 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction