missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP20187, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Description
AP20187 (B/B Homodimerizer) is a cell-permeable ligand used to dimerize FK506-binding protein (FKBP) fusion proteins and initiate biological signaling cascades and gene expression or disrupt protein-protein interactions.
Specifications
Specifications
| Chemical Name or Material | AP20187 |
| CAS | 195514-80-8 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C82H107N5O20 |
| Quantity | 5 mg |
| Synonym | B/B Homodimerizer |
| Solubility Information | Ethanol : 150 mg/mL (101.16 mM; Need ultrasonic) ∣DMSO : ≥ 57 mg/mL (38.44 mM) |
| SMILES | COC1=C(OC)C=C(CC[C@H](C2=CC=CC(OCC(NCC(CN(C)C)CNC(COC3=CC=CC([C@H](OC([C@H]4N(C([C@@H](CC)C5=CC(OC)=C(OC)C(OC)=C5)=O)CCCC4)=O)CCC6=CC(OC)=C(OC)C=C6)=C3)=O)=O)=C2)OC([C@@H]7CCCCN7C([C@H](C8=CC(OC)=C(OC)C(OC)=C8)CC)=O)=O)C=C1 |
| Molecular Weight (g/mol) | 1482.75 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction