missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AP2 beta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598551
This item is not returnable.
View return policy
Description
AP2 beta Polyclonal Antibody for Western Blot, ELISA
This gene encodes a member of the AP-2 family of transcription factors. AP-2 proteins form homo- or hetero-dimers with other AP-2 family members and bind specific DNA sequences. They are thought to stimulate cell proliferation and suppress terminal differentiation of specific cell types during embryonic development. Specific AP-2 family members differ in their expression patterns and binding affinity for different promoters. This protein functions as both a transcriptional activator and repressor. Mutations in this gene result in autosomal dominant Char syndrome, suggesting that this gene functions in the differentiation of neural crest cell derivatives.
Specifications
| AP2 beta | |
| Polyclonal | |
| Unconjugated | |
| Tfap2b | |
| activating enhancer binding protein 2 beta; activating enhancer-binding protein 2 beta; activating enhancer-binding protein 2-beta; AI606113; AP-2(beta); AP-2B; AP2-B; AP2-beta; E130018K07Rik; Tcfap2b; Tfap2b; transcription factor AP-2 beta; transcription factor AP-2 beta (activating enhancer binding protein 2 beta); transcription factor AP-2-beta | |
| Rabbit | |
| Protein G | |
| RUO | |
| 7021 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4.98 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q92481 | |
| Tfap2b | |
| Recombinant Human Transcription factor AP-2-beta protein (111-208AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction