missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AP2 beta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517348
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
This gene encodes a member of the AP-2 family of transcription factors. AP-2 proteins form homo- or hetero-dimers with other AP-2 family members and bind specific DNA sequences. They are thought to stimulate cell proliferation and suppress terminal differentiation of specific cell types during embryonic development. Specific AP-2 family members differ in their expression patterns and binding affinity for different promoters. This protein functions as both a transcriptional activator and repressor. Mutations in this gene result in autosomal dominant Char syndrome, suggesting that this gene functions in the differentiation of neural crest cell derivatives.
Specifications
| AP2 beta | |
| Polyclonal | |
| Unconjugated | |
| Tfap2b | |
| activating enhancer binding protein 2 beta; activating enhancer-binding protein 2 beta; activating enhancer-binding protein 2-beta; AI606113; AP-2(beta); AP-2B; AP2-B; AP2-beta; E130018K07Rik; Tcfap2b; Tfap2b; transcription factor AP-2 beta; transcription factor AP-2 beta (activating enhancer binding protein 2 beta); transcription factor AP-2-beta | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 21419, 301285, 7021 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 20.2 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q61313, Q92481 | |
| Tfap2b | |
| Synthetic peptide corresponding to the sequence of human AP-2β. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction