Learn More
Invitrogen™ AP2 alpha Monoclonal Antibody (K92029_16H5), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | AP2 alpha |
|---|---|
| Clone | K92029_16H5 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30207248
|
Invitrogen™
MA556101 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30204854
|
Invitrogen™
MA555721 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: EDLLHGPHAL SSGLGDLSIH SLPHAIEEVP HVEDPGINIP DQTVIKKGPV SLSKSNSNAV SAIPINKDNL FGGVVNPNEV FCSVPGRLSL LSSTSKYKVT VAEVQRRLSP PECLNASLLG GVLRRAKSKN GGRSLREKLD KIGLNLPAGR RKAANVTLLT SLVEGEAVHL ARDFGYVCET EFPAKAVAEF LNRQHSDPNE QVTRKNMLLA TKQICKEFTD LLAQDRSPLG NSRPNPILEP GIQSCLTHFN LISHGFGSPA VCAAVTALQN YLTEALKAMD KMYLSNNPNS HTDNNAKSSD
AP2 alpha Monoclonal antibody specifically detects AP2 alpha in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| AP2 alpha | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| activating enhancer-binding protein 2 alpha; activating enhancer-binding protein 2-alpha; Activator protein 2; activator protein 2 alpha variant 4; activator protein 2 alpha variant 6; activator protein 2 alpha variant 7; AP2; AP-2; Ap-2 (a); AP-2 transcription factor; AP2alpha; AP-2alpha; AP2-alpha; Ap2tf; BOFS; ERF-1; FLJ51761; OTTHUMP00000214240; OTTHUMP00000214243; Tcfap2a; TFAP2; TFAP2A; transcription factor AP2; transcription factor AP-2 alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); transcription factor AP-2, alpha; transcription factor AP-2-alpha; transcription factor AP-2-alpha-like; transcription factor; AP-2 | |
| Recombinant human TFAP2A (immunogen range is 131-430). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| TFAP2A |
| K92029_16H5 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P05549 | |
| 7020 | |
| IgG2a | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.