Learn More
Invitrogen™ AP2 alpha Monoclonal Antibody (K92029_15F12), Invitrogen™
Description
Sequence of this protein is as follows: EDLLHGPHAL SSGLGDLSIH SLPHAIEEVP HVEDPGINIP DQTVIKKGPV SLSKSNSNAV SAIPINKDNL FGGVVNPNEV FCSVPGRLSL LSSTSKYKVT VAEVQRRLSP PECLNASLLG GVLRRAKSKN GGRSLREKLD KIGLNLPAGR RKAANVTLLT SLVEGEAVHL ARDFGYVCET EFPAKAVAEF LNRQHSDPNE QVTRKNMLLA TKQICKEFTD LLAQDRSPLG NSRPNPILEP GIQSCLTHFN LISHGFGSPA VCAAVTALQN YLTEALKAMD KMYLSNNPNS HTDNNAKSSD
Specifications
Specifications
| Antigen | AP2 alpha |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | K92029_15F12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| Gene | TFAP2A |
| Gene Accession No. | P05549 |
| Gene Alias | activating enhancer-binding protein 2 alpha; activating enhancer-binding protein 2-alpha; Activator protein 2; activator protein 2 alpha variant 4; activator protein 2 alpha variant 6; activator protein 2 alpha variant 7; AP2; AP-2; Ap-2 (a); AP-2 transcription factor; AP2alpha; AP-2alpha; AP2-alpha; Ap2tf; BOFS; ERF-1; FLJ51761; OTTHUMP00000214240; OTTHUMP00000214243; Tcfap2a; TFAP2; TFAP2A; transcription factor AP2; transcription factor AP-2 alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); transcription factor AP-2, alpha; transcription factor AP-2-alpha; transcription factor AP-2-alpha-like; transcription factor; AP-2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.