missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2 alpha Antibody (3B5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
£174.00
Specifications
| Antigen | AP2 alpha |
|---|---|
| Clone | 3B5 |
| Dilution | Western Blot 1:50 - 1:200, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunoprecipitation 1:100 - 1:250, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:50 |
| Applications | Western Blot |
| Classification | Monoclonal |
Beskrivning
AP2 alpha Monoclonal specifically detects AP2 alpha in Human, Mouse, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifikationer
| AP2 alpha | |
| Western Blot 1:50 - 1:200, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunoprecipitation 1:100 - 1:250, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha | |
| TFAP2A | |
| IgG2b | |
| Protein G purified | |
| AP-2 (3B5) |
| 3B5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Transcription Factors and Regulators | |
| P05549 | |
| 7020 | |
| N-terminus of human AP2a. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 48 kDa |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel