missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AP1S1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-28986
This item is not returnable.
View return policy
Description
AP1S1 Polyclonal antibody specifically detects AP1S1 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| AP1S1 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| adaptor-related protein complex 1, sigma 1 subunit, AP19HA1 19 kDa subunit, CLAPS1FLJ92436, Clathrin assembly protein complex 1 sigma-1A small chain, Clathrin coat assembly protein AP19, Golgi adaptor HA1/AP1 adaptin sigma-1A subunit, Sigma 1a subunit of AP-1 clathrin, SIGMA1A, sigma1A subunit of AP-1 clathrin adaptor complex, Sigma1A-adaptin, Sigma-adaptin 1A, small 1 (19kD) | |
| Between 108 and 158 | |
| 100 μg | |
| Immunology, Signal Transduction | |
| 1174 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction