missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP1S1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92032-0.02ml
This item is not returnable.
View return policy
Description
AP1S1 Polyclonal antibody specifically detects AP1S1 in Human samples. It is validated for Western Blot
Specifications
| AP1S1 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| adaptor-related protein complex 1, sigma 1 subunit, AP19HA1 19 kDa subunit, CLAPS1FLJ92436, Clathrin assembly protein complex 1 sigma-1A small chain, Clathrin coat assembly protein AP19, Golgi adaptor HA1/AP1 adaptin sigma-1A subunit, Sigma 1a subunit of AP-1 clathrin, SIGMA1A, sigma1A subunit of AP-1 clathrin adaptor complex, Sigma1A-adaptin, Sigma-adaptin 1A, small 1 (19kD) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human AP1S1 (NP_001274.1). MMRFMLLFSRQGKLRLQKWYLATSDKERKKMVRELMQVVLARKPKMCSFLEWRDLKVVYKRYASLYFCCAIEGQDNELITLELIHRYVELLDKYFGSVCELDIIFNFEKAYFILDEFLMG | |
| 0.02 mL | |
| Immunology, Signal Transduction | |
| 1174 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction