Learn More
Invitrogen™ AP1M2 Polyclonal Antibody

Description
Immunogen sequence: IKYKKNEVFI DVIESVNLLV NANGSVLLSE IVGTIKLKVF LSGMPELRLG LNDRVLFELT GRSKNKSVEL EDVKFHQCVR LSRFDNDRTI SFIPPDGDFE LMSYRLSTQV KPLIWIESVI EKFSHSRVEI MVKAKGQFKK QSVANGVEIS VPVPSDADSP RFKTSVGSAK YVPERNVVIW SIKSFPGGKE YLMRAHFGLP SVEKEEVEGR PPIGVKFEIP YFTVSGIQVR YMKIIEKSGY QALPWVRYIT QSGDYQLRTS; Positive Samples: 22Rv1, BT-474, HepG2, Mouse kidney, Mouse lung, Mouse small intestine; Cellular Location: Cytoplasmic side, Cytoplasmic vesicle, Golgi apparatus, Peripheral membrane protein, clathrin-coated vesicle membrane.
Specifications
Specifications
| Antigen | AP1M2 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.33 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | AP1M2 |
| Gene Accession No. | Q9WVP1, Q9Y6Q5 |
| Gene Alias | [m]1B; adaptor protein complex AP-1 mu-2 subunit; adaptor protein complex AP-1 subunit mu-2; adaptor protein complex AP-1, mu 2 subunit; adaptor related protein complex 1 mu 2 subunit; adaptor-related protein complex 1 mu 2 subunit; adaptor-related protein complex 1 mu-2 subunit; adaptor-related protein complex 1 subunit mu-2; adaptor-related protein complex 1, mu 2 subunit; adaptor-related protein complex AP-1, mu 2 subunit; AP-1 complex subunit mu-2; Ap1m2; AP1-mu2; AP-mu chain family member mu1B; clathrin assembly protein complex 1 medium chain 2; clathrin assembly protein complex 1 mu-2 medium chain 2; clathrin coat assembly protein AP47 2; clathrin coat associated protein AP47 2; clathrin-associated adaptor medium chain mu2; D9Ertd818e; golgi adaptor AP-1 47 kDa protein; golgi adaptor HA1/AP1 adaptin mu-2 subunit; HA1 47 kDa subunit 2; HSMU1B; MU1B; MU-1B; Mu1B-adaptin; mu2; Mu-adaptin 2; RGD1561490 |
| Gene Symbols | AP1M2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.