missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP1M1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12112-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is the medium chain of the trans-Golgi network clathrin-associated protein complex AP-1. The other components of this complex are beta-prime-adaptin, gamma-adaptin, and the small chain AP1S1. This complex is located at the Golgi vesicle and links clathrin to receptors in coated vesicles. These vesicles are involved in endocytosis and Golgi processing. Alternatively spliced transcript variants encoding distinct protein isoforms have been found for this gene.Specifications
| AP1M1 | |
| Polyclonal | |
| Unconjugated | |
| AP1M1 | |
| AP 1 complex subunit mu 1, AP mu chain family member mu1A, AP1M1, AP47, CLAPM2, CLTNM, MU 1A, Mu adaptin 1, Mu1A adaptin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11767, 306332, 8907 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P35585, Q32Q06, Q9BXS5 | |
| AP1M1 | |
| AP1M1 Fusion Protein Ag2757 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction