Learn More
Invitrogen™ AP1M1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587656
Description
Immunogen sequence: MSASAVYVLD LKGKVLICRN YRGDVDMSEV EHFMPILMEK EEEGMLSPIL AHGGVRFMWI KHNNLYLVAT SKKNACVSLV FSFLYKVVQV FSEYFKELEE ESIRDNFVII YELLDELMDF GYPQTTDSKI LQEYITQEGH KLETGAPRPP ATVTNAVSWR; Positive Samples: HeLa, Jurkat, HepG2, HL-60, HT-1080, Mouse brain, Mouse kidney, Mouse liver, Rat testis; Cellular Location: Cytoplasmic side, Cytoplasmic vesicle, Golgi apparatus, Peripheral membrane protein, clathrin-coated vesicle membrane.
The protein encoded by this gene is the medium chain of the trans-Golgi network clathrin-associated protein complex AP-1. The other components of this complex are beta-prime-adaptin, gamma-adaptin, and the small chain AP1S1. This complex is located at the Golgi vesicle and links clathrin to receptors in coated vesicles. These vesicles are involved in endocytosis and Golgi processing. Alternatively spliced transcript variants encoding distinct protein isoforms have been found for this gene.
Specifications
| AP1M1 | |
| Polyclonal | |
| Unconjugated | |
| AP1M1 | |
| [m]1A; AA408894; adapter-related protein complex 1 subunit mu-1; adaptor protein complex AP-1 mu-1 subunit; adaptor protein complex AP-1 subunit mu-1; adaptor related protein complex 1 mu 1 subunit; adaptor-related protein complex 1 mu 1 subunit; adaptor-related protein complex 1 mu-1 subunit; adaptor-related protein complex 1 subunit mu-1; adaptor-related protein complex 1, mu 1 subunit; adaptor-related protein complex AP-1, mu subunit 1; Adtm1A; AP-1 complex subunit mu-1; Ap1m1; ap1m1 {ECO:0000312; AP47; AP-mu chain family member mu1A; CLAPM2; clathrin adaptor protein AP47; clathrin assembly protein complex 1 medium chain 1; clathrin assembly protein complex 1 mu-1 medium chain 1; clathrin assembly protein complex 1, medium chain; clathrin assembly protein complex AP1, mu subunit; clathrin coat assembly protein AP47; Clathrin coat-associated protein AP47; Cltnm; golgi adaptor AP-1 47 kDa protein; golgi adaptor HA1/AP1 adaptin mu-1 subunit; HA1 47 kDa subunit; mu1A; MU-1A; mu1A-adaptin; Mu-adaptin 1; RGD:1307653} | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11767, 306332, 8907 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35585, Q32Q06, Q9BXS5 | |
| AP1M1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human AP1M1 (NP_1158821). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.