missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP1M1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £359.00
Specifications
| Antigen | AP1M1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627251
|
Novus Biologicals
NBP2-92000-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613211
|
Novus Biologicals
NBP2-92000-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AP1M1 Polyclonal antibody specifically detects AP1M1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| AP1M1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Immunology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 8907 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Adaptor protein complex AP-1 mu-1 subunit, Adaptor-related protein complex 1 mu-1 subunit, adaptor-related protein complex 1, mu 1 subunit, AP47MU-1A, AP-mu chain family member mu1A, CLAPM2AP-1 complex subunit mu-1, clathrin adaptor protein AP47, Clathrin assembly protein complex 1 medium chain 1, clathrin assembly protein complex 1, medium chain, clathrin assembly protein complex AP1, mu subunit, Clathrin coat assembly protein AP47, Clathrin coat-associated protein AP47, CLTNM, golgi adaptor AP-1 47 kDa protein, Golgi adaptor HA1/AP1 adaptin mu-1 subunit, HA1 47 kDa subunit, Mu1A-adaptin, Mu-adaptin 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human AP1M1 (NP_115882.1). MSASAVYVLDLKGKVLICRNYRGDVDMSEVEHFMPILMEKEEEGMLSPILAHGGVRFMWIKHNNLYLVATSKKNACVSLVFSFLYKVVQVFSEYFKELEEESIRDNFVIIYELLDELMDFGYPQTTDSKILQEYITQEGHKLETGAPRPPATVTNAVSWR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title