missing translation for 'onlineSavingsMsg'
Learn More

AP1867, MedChemExpress

Product Code. 16465626
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465626 100 mg 100mg
16431097 50 mg 50mg
16424838 10 mg 10mg
16407587 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16465626 Supplier Medchem Express Supplier No. HY114434
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AP1867 is a synthetic FKBP12F36V-directed ligand.

AP1867 is a synthetic FKBP12F36V-directed ligand.

Specifications

Chemical Name or Material AP1867
CAS 195514-23-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C38H47NO11
Quantity 100 mg
Solubility Information DMSO : ≥ 125 mg/mL (180.17 mM)
SMILES COC1=CC=C(CC[C@@H](OC([C@@H]2CCCCN2C([C@H](C3=CC(OC)=C(OC)C(OC)=C3)CC)=O)=O)C4=CC=CC(OCC(O)=O)=C4)C=C1OC
Molecular Weight (g/mol) 693.78
Formula Weight 693.78
Percent Purity 98.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.