missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP180 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38325-20ul
This item is not returnable.
View return policy
Description
AP180 Polyclonal antibody specifically detects AP180 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| AP180 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| 91 kDa synaptosomal-associated protein, AP180, CALM, clathrin coat assembly protein AP180, DKFZp781O0519, KIAA0656synaptosomal-associated protein, 91 kDa (mouse) homolog, Phosphoprotein F1-20, synaptosomal-associated protein, 91kDa homolog (mouse) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 321-600 of human AP180 (NP_001229723.1).,, Sequence:, AVPVSTSKPSSDLLDLQPDFSSGGAAAAAAPAPPPPAGGATAWGGFGGSFMAPSPSPVTPAQNNLLQPNFEAAFGTTPSTSSSSSFDPSVFDGLGDLLMPTMAPAGQPAPVSMVPPSPAMAASKALGSDLDSSLASLVGNLGISGTTTKKGDLQWNAGEKKLTGGANWQPKVAPATWSAGVPPSAPLQGAVPPTSSVPPVAGAPSVGQPGAGFGMPPAGTGMPMMPQQPVMFAQPMMRPPFGAAAVPGTQLSPSPTPASQSPKKPPAKDPLADLNIKDFL | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9892 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction