missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AP Enhancer Solution
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Description
AP Enhancer Solution, 10 mL is an aqueous buffered solution and an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays. It is an enhancer solution for alkaline phosphatase.
Specifications
Specifications
| Content And Storage | Store at 2°C to 8°C |
| Product Type | AP Enhancer Solution |
| Quantity | 10 mL |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction