missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AP C5
GREENER_CHOICE

Product Code. 18768626
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18768626

Brand: Tocris Bioscience™ 6626/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective PKG2 inhibitor

AP C5 is a potent and selective protein kinase G type II (PKG2) inhibitor (pIC50 = 7.2). Displays >20-fold selectivity for PKG2 over PKG1 and PKA (pIC50 values are 4.6 and 4.8 for PKG1 and PKA, respectively). Blocks 8-pCPT-cGMP-dependent VASP phosphorylation in intestinal organoids. Also attenuates toxin-stimulated anion secretory responses in mouse intestinal tissue.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[4-(1H-Imidazol-1-yl)phenyl]-N-2-propyn-1-yl-2-pyrimidinamine
CAS 2234272-10-5
Quantity 10 mg
Molecular Formula C16H13N5
Formula Weight 275.31
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.