missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP-2 gamma Antibody (3A11A5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £437.00
Specifications
| Antigen | AP-2 gamma |
|---|---|
| Clone | 3A11A5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18084876
|
Novus Biologicals
NBP2-61859 |
0.1 mL |
£437.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627887
|
Novus Biologicals
NBP2-61859-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AP-2 gamma Monoclonal specifically detects AP-2 gamma in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| AP-2 gamma | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| 7022 | |
| Purified recombinant fragment of human TFAP2C (AA: 341-450) expressed in E. Coli. | |
| Primary | |
| 49.2 kDa |
| 3A11A5 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| Activating enhancer-binding protein 2 gamma, AP2-GAMMA, ERF1, estrogen receptor factor 1, hAP-2g, TFAP2G, transcription factor AP-2 gamma, transcription factor AP-2 gamma (activating enhancer binding protein 2 gamma), transcription factor AP-2 gamma (activating enhancer-binding protein 2 gamma), Transcription factor ERF-1 | |
| TFAP2C | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title