missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP-2 beta/TFAP2B Antibody (6F7G1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | AP-2 beta/TFAP2B |
|---|---|
| Clone | 6F7G1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18243831
|
Novus Biologicals
NBP2-61858 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685678
|
Novus Biologicals
NBP2-61858-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AP-2 beta/TFAP2B Monoclonal specifically detects AP-2 beta/TFAP2B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| AP-2 beta/TFAP2B | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| activating enhancer binding protein 2 beta, Activating enhancer-binding protein 2-beta, AP-2B, AP2-B, AP2-beta, CHAR, MGC21381, transcription factor AP-2 beta (activating enhancer binding protein 2 beta), transcription factor AP-2 beta (activating enhancer-binding protein 2 beta), transcription factor AP-2-beta | |
| TFAP2B | |
| IgG2b | |
| Protein G purified |
| 6F7G1 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Neuroscience | |
| 7021 | |
| Purified recombinant fragment of human TFAP2B (AA: 84-193) expressed in E. Coli. | |
| Primary | |
| 50.5 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title