missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AP 14145 hydrochloride
GREENER_CHOICE

Product Code. 18799696
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18799696 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18799696 Supplier Tocris Bioscience™ Supplier No. undefined

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Negative allosteric modulator of KCa2 (SK) channels

AP 14145 hydrochloride is a KCa2 (small conductance Ca2+-activated potassium) channel negative allosteric modulator (IC50 = 1.1 μM). Increases the EC50 of Ca2+ on KCa2.3 channels by ~3-fold. Prolongs atrial effective refractory period (AERP) in rats. Reduces atrial fibrillation (AF) duration and prolongs atrial refractoriness without affecting ventricular refractory period in an animal AF model.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[2-[[(1R)-1-[3-(Trifluoromethyl)phenyl]ethyl]amino]-1H-benzimidazol-7-yl]acetamide hydrochloride
CAS 2387505-59-9
Quantity 5 mg
Molecular Formula C18H17F3N4O.HCl
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.