missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AOX1 Recombinant Rabbit Monoclonal Antibody (PSH0-17)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550783
This item is not returnable.
View return policy
Description
Positive control: Rat liver tissue lysates, rat liver tissue, rat adrenal gland tissue, human kidney tissue, mouse testis tissue, Hela. Predicted band size: 148 kDa Subcellular Location: Cytoplasm.
AOX1 (Aldehyde oxidase 1) is a 1338 amino acid cytoplasmic protein that catalyzes the oxidation of a variety of aldehydes, leading to the production of hydrogen peroxide. Under certain conditions, AOX1 can catalyze the formation of the superoxide free radical. Defects in oxygen radical metabolism have been linked to the pathogenesis of amyotrophic lateral sclerosis (ALS), an autosomal dominant neurodegenerative disorder characterized by the death of motor neurons in the spinal cord, brain and brainstem. Significantly, AOX1 is highly expressed in the ventral horn of the spinal cord and the gene that encodes AOX1 is located in a chromosomal region that is frequently found to be implicated in ALS2. This evidence suggests that AOX1 is a candidate gene for ALS2.
Specifications
| AOX1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O54754, Q06278, Q9Z0U5 | |
| AOX1 | |
| Synthetic peptide within human AOX1 aa 1301-1338/1338. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| PSH0-17 | |
| Unconjugated | |
| AOX1 | |
| AI196512; AI255253; Aldehyde oxidase; aldehyde oxidase (female form); aldehyde oxidase 1; aldehyde oxidase 2; Ao; AOH1; Aox1; Aox-1; Aox2; Aox-2; Azaheterocycle hydroxylase; azaheterocycle hydroxylase 1; Moro; Retinal oxidase; Ro | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11761, 316, 54349 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction