missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AOX1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16607894
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16607894 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16607894 Supplier Invitrogen™ Supplier No. PA5101413

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AOX1.

AOX1 (Aldehyde oxidase 1) is a 1338 amino acid cytoplasmic protein that catalyzes the oxidation of a variety of aldehydes, leading to the production of hydrogen peroxide. Under certain conditions, AOX1 can catalyze the formation of the superoxide free radical. Defects in oxygen radical metabolism have been linked to the pathogenesis of amyotrophic lateral sclerosis (ALS), an autosomal dominant neurodegenerative disorder characterized by the death of motor neurons in the spinal cord, brain and brainstem. Significantly, AOX1 is highly expressed in the ventral horn of the spinal cord and the gene that encodes AOX1 is located in a chromosomal region that is frequently found to be implicated in ALS2. This evidence suggests that AOX1 is a candidate gene for ALS2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AOX1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AOX1
Gene Accession No. O54754, Q06278, Q9Z0U5
Gene Alias AI196512; AI255253; Aldehyde oxidase; aldehyde oxidase (female form); aldehyde oxidase 1; aldehyde oxidase 2; Ao; AOH1; Aox1; Aox-1; Aox2; Aox-2; Azaheterocycle hydroxylase; azaheterocycle hydroxylase 1; Moro; Retinal oxidase; Ro
Gene Symbols AOX1
Host Species Rabbit
Immunogen A synthesized peptide derived from human AOX1(Accession Q06278), corresponding to amino acid residues K525-H575.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11761, 316, 54349
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.