missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AOX1 Polyclonal Antibody

Description
Positive Control: Mouse liver tissue lysate, rat liver tissue lysate, human liver carcinoma tissue, human liver tissue, Hela. Subcellular Location: Cytoplasm.
AOX1 (Aldehyde oxidase 1) is a 1338 amino acid cytoplasmic protein that catalyzes the oxidation of a variety of aldehydes, leading to the production of hydrogen peroxide. Under certain conditions, AOX1 can catalyze the formation of the superoxide free radical. Defects in oxygen radical metabolism have been linked to the pathogenesis of amyotrophic lateral sclerosis (ALS), an autosomal dominant neurodegenerative disorder characterized by the death of motor neurons in the spinal cord, brain and brainstem. Significantly, AOX1 is highly expressed in the ventral horn of the spinal cord and the gene that encodes AOX1 is located in a chromosomal region that is frequently found to be implicated in ALS2. This evidence suggests that AOX1 is a candidate gene for ALS2.
Specifications
Specifications
| Antigen | AOX1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | AOX1 |
| Gene Accession No. | O54754, Q06278, Q9Z0U5 |
| Gene Alias | AI196512; AI255253; Aldehyde oxidase; aldehyde oxidase (female form); aldehyde oxidase 1; aldehyde oxidase 2; Ao; AOH1; Aox1; Aox-1; Aox2; Aox-2; Azaheterocycle hydroxylase; azaheterocycle hydroxylase 1; Moro; Retinal oxidase; Ro |
| Gene Symbols | AOX1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?