missing translation for 'onlineSavingsMsg'
Learn More

AOF1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18424056 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424056 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424056 Supplier Novus Biologicals Supplier No. NBP281895MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AOF1 Polyclonal antibody specifically detects AOF1 in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen AOF1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias AOF1, bA204B7.3, C6orf193, dJ298J15.2, DKFZp686I0412, flavin containing 1, flavin-containing amine oxidase domain-containing protein 1, FLJ33898, FLJ34109, FLJ43328, LSD2, lysine (K)-specific demethylase 1B, lysine-specific histone demethylase 1B, lysine-specific histone demethylase 2
Host Species Rabbit
Immunogen AOF1 antibody was raised against a 17 amino acid peptide near the amino terminus of human AOF1. The immunogen is located within amino acids 80 - 130 of AOF1. Amino Acid Squence: FDHYYRSHKDGYDKYT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221656
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.