missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anza™ T4 DNA Ligase Master Mix

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

Brand:  Invitrogen™ IVGN2108

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15501620

  • £716.00 / Each
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Anza™ T4 DNA Ligase Master Mix

Description

Description

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One Ligase for both cohesive and blunt end ligations
• 4X conc. enables higher volume of dilute DNA in reaction mixture
• Ligation complete in 15 minutes
• Convenient Master Mix format reduces pipetting

Certifications

CoA

Specifications

Specifications

4X
Approved for shipment on Wet or Dry Ice
Elution Buffer, 1X Anza Buffer
T4 DNA Ligase Master Mix
1 mL Anza T4 DNA Ligase Master Mix

Store at -5 to -30°C.
T4 DNA Ligase
200 Reactions
Anza™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.