missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anza™ 89 Mva1269I

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

Brand:  Invitrogen™ IVGN0894

Additional Details : Weight : 0.01000kg

Product Code. 15671952

  • £90.00 / 400 units
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer

Description

Description

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • One buffer for all restriction enzymes
  • One digestion protocol for all DNA types
  • Complete digestion in 15 min
  • Overnight digestion without star activity

Certifications

CoA

Specifications

Specifications

40 μL Anza 89 Mva1269I
500 μL Anza Buffer (10X)
500 μL Anza Red Buffer (10X)

Store at -20°C.
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Anza™ 10X Red Buffer, Anza™ 10X Buffer
37°C
Restriction Enzyme Cloning System
Anza™
Approved for shipment on Wet or Dry Ice
Mva1269I
Yes
Yes
Traditional Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.