missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Anza™ 60 Kpn2I
Click to view available options
Quantity:
800 units
Unit Size:
800 units
Description
Includes
Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
- One buffer for all restriction enzymes
- One digestion protocol for all DNA types
- Complete digestion in 15 min
- Overnight digestion without star activity
Certifications
CoA
Specifications
Specifications
| Content And Storage | 40 μL Anza 60 Kpn2I 500 μL Anza Buffer (10X) 500 μL Anza Red Buffer (10X) Store at -20°C. |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
| Methylation Sensitivity | CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive |
| Enzyme | Kpn2I |
| Compatible Buffer | Anza™ 10X Red Buffer, Anza™ 10X Buffer |
| Sensitive to Heat Inactivation | Yes |
| Optimal Reaction Temperature | 37°C |
| Type IIS RE | No |
| Quantity | 800 units |
| Product Type | Restriction Enzyme Cloning System |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction