missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Anza™ 5 BamHI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
£64.50 - £168.00
Specifications
Shipping Condition | Approved for shipment on Wet or Dry Ice |
---|---|
Methylation Sensitivity | Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive |
Enzyme | BamH I |
Compatible Buffer | Anza™ 10X Red Buffer, Anza™ 10X Buffer |
Sensitive to Heat Inactivation | Yes |
Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15515680
|
Invitrogen™
IVGN0058 |
24,000 U |
£168.00
1 set |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15505680
|
Invitrogen™
IVGN0056 |
8000 U |
£64.50
1 set |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity
Certifications
CoA
Specifications
Approved for shipment on Wet or Dry Ice | |
BamH I | |
Yes | |
No | |
Traditional Cloning |
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive | |
Anza™ 10X Red Buffer, Anza™ 10X Buffer | |
37°C | |
Restriction Enzyme Cloning System | |
Anza™ |
For Research Use Only. Not for use in diagnostic procedures.