missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anza™ 22 SmaI

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

Brand:  Invitrogen™ IVGN0228

Code : NEW

Additional Details : Weight : 0.40000kg

 View more versions of this product

Product Code. 15535710

  • £356.00 / 1 set
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer

Description

Description

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity

Certifications

CoA

Specifications

Specifications

1.6 mL Anza 22 SmaI
2 mL Anza Buffer (10X)
2 mL Anza Red Buffer (10X)

Store at -20°C.
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Anza™ 10X Red Buffer, Anza™ 10X Buffer
37°C
8000 U
Traditional Cloning
Approved for shipment on Wet or Dry Ice
Sma I
Yes
No
Restriction Enzyme Cloning System
Anza™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.