Learn More
Invitrogen™ ANXA8L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596743
Description
Immunogen sequence: MAWWKAWIEQ EGVTVKSSSH FNPDPDAETL YKAMKGIGVG SQLLSHQAAA FAFPSSALTS VSPWGQQGHL CCNPAGTNEQ AIIDVLTKRS NTQRQQIAKS FKAQFGKDLT ETLKSELSGK FERLIVALMY PPYRYEAKEL HDAMKGSRDD VSSFVDPALA LQDAQDLYAA GEKIRGTDEM KFITILCTRS ATHLLRVKCT QNLHSYFAER LYYAMKGAGT RDGTLIRNIV SRSEIDLNLI KCHFKKMYGK TLSSMIMEDT SGDYKNALLS LVGSDP; Positive Samples: A-549, HeLa, Mouse lung, Mouse ovary.
This gene encodes a member of the annexin family of evolutionarily conserved Ca2+ and phospholipid binding proteins. The encoded protein may function as an anticoagulant that indirectly inhibits the thromboplastin-specific complex. Overexpression of this gene has been associated with acute myelocytic leukemia. A highly similar duplicated copy of this gene is found in close proximity on the long arm of chromosome 10.
Specifications
| ANXA8L2 | |
| Polyclonal | |
| Unconjugated | |
| ANXA8L1 | |
| AI466995; annexin 8; annexin A8; annexin A8L2; annexin A8-like 1; annexin A8-like protein 1; annexin A8-like protein 2; annexin VIII; Annexin-8; Anx8; ANXA8; ANXA8L1; ANXA8L2; bA145E20.2; LOC100052045; SPT7 like, STAGA complex gamma subunit; SPT7 like, STAGA complex subunit gamma; SPTF-associated factor 65 gamma; STAGA complex 65 subunit gamma; suppressor of Ty 7-like; SUPT7L; Unknown (protein for MGC:137593); VAC-beta; Vascular anticoagulant-beta | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 728113 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5VT79 | |
| ANXA8L1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-276 of human ANXA8L2 (NP_0012658531). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.