missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Antp Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569514
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Fruit fly:100%;.
Antp gene ontology annotations related to this gene include multicellular organism development.
Specifications
| Antp | |
| Polyclonal | |
| Unconjugated | |
| Antp | |
| 3.4; Ant; ANTC; ANT-C; antenapedia; antennapaedia; antennapedia; antennapedia P1; antennapedia P2; antennapedia protein; antp; ANT-P; Antp P1; Antp P2; AntP1; Antp-PD; Antp-PE; Antp-PF; Antp-PG; Antp-PH; Antp-PI; Antp-PJ; Antp-PK; Antp-PL; Antp-PM; Antp-PN; Antp-PO; atennapedia; Aus; BG:DS07700.1; CG1028; CG1028-PD; CG1028-PE; CG1028-PF; CG1028-PG; CG1028-PH; CG1028-PI; CG1028-PJ; CG1028-PK; CG1028-PL; CG1028-PM; CG1028-PN; CG1028-PO; Dm-Antennapedia; DmAntp; DMANTPE1; Dmel\CG1028; Dmel_CG1028; DRO15DC96Z; Homeotic protein antennapedia; Hu; humeral; l(3)84Ba; LD33666p; nasobemia; Ns; Scx; Sex comb extra; Sex combs extra | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 40835 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P02833 | |
| Antp | |
| synthetic peptide corresponding to a region of fruit fly Antp (aa 151-200). | |
| 100 μL | |
| Primary | |
| Drosophila | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction