missing translation for 'onlineSavingsMsg'
Learn More

Antitumor agent-3, MedChemExpress

Product Code. 16489477
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489477 5 mg 5mg
16427016 100 mg 100mg
16426938 10 mg 10mg
16402997 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489477 Supplier Medchem Express Supplier No. HY15961
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Antitumor agent-3 is a potent compound which comprises a new imidazopyridine having excellent antitumor activity as an active ingredient.

Antitumor agent-3 is a potent compound which comprises a new imidazopyridine having excellent antitumor activity as an active ingredient.

Specifications

Chemical Name or Material Antitumor agent-3
CAS 420126-30-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H10F6N4S
Quantity 5 mg
Solubility Information DMSO : ≥ 59 mg/mL (137.74 mM)
SMILES FC(C1=CC=C(NC2=NC(C3=C(C(F)(F)F)N=C4C=CC=CN43)=CS2)C=C1)(F)F
Molecular Weight (g/mol) 428.35
Formula Weight 428.35
Percent Purity 99.71%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.