Learn More
Mettler Toledo™ Antistatic Kit, Universal
Product Code. 10533744 Shop All Mettler Toledo Products
Change view
Click to view available options
For Use With (Equipment):
Laboratory Balance
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10533744 Laboratory Balance Each
1 options
Product Code. 10533744 Supplier Mettler Toledo™ Supplier No. 11107767
Completely remove electrostatic charge from your samples and tare containers and avoid distortion of your weighing results. Mettler Toledo™ Antistatic Kit, Universal enables fast dosing processes and substantially reduces the risk of contamination.
  • Large electrode for discharging all kinds of samples and tare containers with dimensions of 15x25cm
  • Improves weighing accuracy
  • Enables fast dosing
  • Reduces contamination risk
  • Can be freely placed next to your balance
  • Includes universal power supply
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Laboratory Balance
For Use With (Application) Discharging electrostatic charge
Includes Universal power supply, Large U-Electrode
Size Large