missing translation for 'onlineSavingsMsg'
Learn More

Antipyrine, 98%

Product Code. 11478953
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478953 100 g 100g
11488953 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478953 Supplier Thermo Scientific Alfa Aesar Supplier No. A11089.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 60-80-0 | C11H12N2O | 188.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-80-0
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD00003146
InChI Key VEQOALNAAJBPNY-UHFFFAOYSA-N
Synonym antipyrine, phenazone, antipyrin, phenazon, analgesine, anodynine, anodynin, azophen, fenazone, 2,3-dimethyl-1-phenyl-5-pyrazolone
PubChem CID 2206
ChEBI CHEBI:31225
IUPAC Name 1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=CC(=O)N(N1C)C2=CC=CC=C2

Specifications

Melting Point 111°C to 114°C
Quantity 100 g
Beilstein 157775
Merck Index 14,716
Formula Weight 188.23
Percent Purity 98%
Chemical Name or Material Antipyrine
compliance-icons
Product Identifier
  • Antipyrine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Antipyrine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.