missing translation for 'onlineSavingsMsg'
Learn More

Antipain dihydrochloride

Product Code. 15465859
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 15465859

Brand: Thermo Scientific Alfa Aesar J63680.#0

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 37682-72-7 | C27H46Cl2N10O6 | 677.63 g/mol

Antipain inhibits trypsin, papain, and catherpsins A and B (a reversible inhibitor of cysteine and serine proteases). It is used to evaluate the role of proteases in cell transformations. It is used to help identify new proteases.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Antipain inhibits trypsin, papain, and catherpsins A and B (a reversible inhibitor of cysteine and serine proteases). It is used to evaluate the role of proteases in cell transformations. It is used to help identify new proteases.
TRUSTED_SUSTAINABILITY

Specifications

CAS 37682-72-7
Color White to Yellow
Molecular Formula C27H46Cl2N10O6
MDL Number MFCD00135957
Quantity 5 mg
Synonym antipain dihydrochloride
Solubility Information 0.01M in water
InChI Key GXQPAKIVOCLCHD-BXLPLHKWSA-N
SMILES [H+].[H+].[Cl-].[Cl-].CC(C)[C@H](NC(CCCN=C(N)N)C=O)C(=O)NC(=O)[C@H](CCCN=C(N)N)NC(=O)NC(CC1=CC=CC=C1)C(O)=O
IUPAC Name 2-[[5-(diaminomethylideneamino)-1-[[1-[[5-(diaminomethylideneamino)-1-oxopentan-2-yl]amino]-3-methyl-1-oxobutan-2-yl]amino]-1-oxopentan-2-yl]carbamoylamino]-3-phenylpropanoic acid;dihydrochloride
Molecular Weight (g/mol) 677.63
PubChem CID 24847649
Formula Weight 677.6
Concentration or Composition (by Analyte or Components) 0.01M in water
Physical Form Powder
Chemical Name or Material Antipain dihydrochloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.