missing translation for 'onlineSavingsMsg'
Learn More

Antineoplaston A10, MedChemExpress

Product Code. 16495398
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495398 10 mg 10mg
16451587 50 mg 50mg
16448117 5 mg 5mg
16436006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495398 Supplier Medchem Express Supplier No. HY128553

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Antineoplaston A10, a naturally occurring substance in human body, is a Ras inhibitor potentially for the treatment of glioma, lymphoma, astrocytoma and breast cancer.

Antineoplaston A10, a naturally occurring substance in human body, is a Ras inhibitor potentially for the treatment of glioma, lymphoma, astrocytoma and breast cancer.

Specifications

Chemical Name or Material Antineoplaston A10
CAS 91531-30-5
Color White
Purity Grade Notes Research
Molecular Formula C13H14N2O3
Quantity 10 mg
Solubility Information DMSO : 250 mg/mL (1015.19 mM; Need ultrasonic)
SMILES O=C(N[C@@H](CC1)C(NC1=O)=O)CC2=CC=CC=C2
Molecular Weight (g/mol) 246.26
Formula Weight 246.26
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.