missing translation for 'onlineSavingsMsg'
Learn More

Antimycin A

Product Code. 15495729
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495729 5 mg 5mg
15405739 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495729 Supplier Thermo Scientific Alfa Aesar Supplier No. J63522.#0
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1397-94-0 | C28H40N2O9 | 548.633 g/mol

Antimycin A is used to study the specific sites of reactive oxygen species production in mitochondria isolated from skeletal muscle of chronic obstructive pulmonary disease patients, and its relationship with local oxidative stress induced by exercise.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimycin A is used to study the specific sites of reactive oxygen species production in mitochondria isolated from skeletal muscle of chronic obstructive pulmonary disease patients, and its relationship with local oxidative stress induced by exercise.

Solubility
Soluble in 100% ethanol. Insoluble in water.

Notes
Isolated from Streptomyces sp.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1397-94-0
Molecular Formula C28H40N2O9
Molecular Weight (g/mol) 548.633
MDL Number MFCD01779723
InChI Key UIFFUZWRFRDZJC-RBVQMQRASA-N
PubChem CID 16218979
IUPAC Name [(2R,3S,6S,7R)-3-[(3-formamido-2-hydroxybenzoyl)amino]-8-hexyl-2,6-dimethyl-4,9-dioxo-1,5-dioxonan-7-yl] 3-methylbutanoate
SMILES CCCCCCC1C(C(OC(=O)C(C(OC1=O)C)NC(=O)C2=C(C(=CC=C2)NC=O)O)C)OC(=O)CC(C)C

Specifications

Type Antimycin A
Physical Form Powder
Melting Point 149°C to 150°C
Color White to Yellow
Quantity 5 mg
Sensitivity Light sensitive
UN Number UN3462
Merck Index 14,714
Solubility Information Soluble in 100% ethanol. Insoluble in water.
Formula Weight 548.63
Chemical Name or Material Antimycin A
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Antimycin A
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contain: Antimycin A

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.