missing translation for 'onlineSavingsMsg'
Learn More

Antimony(V) chloride, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049660
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
7647-18-9
Cl5H8Sb
307.07
MFCD00011213
QFJJIKGZKLCYFT-UHFFFAOYSA-N
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
24294
pentachloro-$l^{5}-stibane
Cl.Cl.Cl.Cl.Cl.[SbH3]
This item is not returnable. View return policy
7647-18-9
Cl5H8Sb
307.07
MFCD00011213
QFJJIKGZKLCYFT-UHFFFAOYSA-N
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
24294
pentachloro-$l^{5}-stibane
Cl.Cl.Cl.Cl.Cl.[SbH3]

Antimony(V) chloride is used as a catalyst and an analytical reagent for testing alkaloids and cesium. It is also used as a polymerization catalyst as well as involved in the chlorination of organic compounds. It acts as a Lewis acid and a strong oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 79°C (22mmHg)
Melting Point 2.8°C
Color Red-Yellow
Physical Form Liquid
Quantity 25 g
Linear Formula SbCl5
UN Number UN1730
Merck Index 14,695
Solubility Information Miscible with chloroform,hydrochloric acid,tartaric acid,carbon disulfide and carbon tetrachloride.
Formula Weight 299.02
Percent Purity 99.997%
Odor Pungent
Refractive Index 1.601
Sensitivity Moisture sensitive
Density 2.336 g/mL
Assay (metals basis)
Chemical Name or Material Antimony(V) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'