missing translation for 'onlineSavingsMsg'
Learn More

Antimony trifluoride, 98%, ACROS Organics™

Brand:  Acros Organics 191241000

Code : 29

Additional Details : CAS Number : 7783-56-4 Weight : 0.15000kg Transport : UN number : 2923 Chem class : 8 Pack group : II


Packaging Plastic bottle
Quantity 100g

 View more versions of this product

Product Code. 10002450

  • £24.41 / 100g

In stock, ready to ship: 4

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

7783-56-4
178.75
GUNJVIDCYZYFGV-UHFFFAOYSA-K
24554
F[Sb](F)F
F3Sb
MFCD00011218
antimony trifluoride, trifluorostibine, antimony fluoride, antimony iii fluoride, stibine, trifluoro, trifluoroantimony, antimoine fluorure, antimonous fluoride, antimoine fluorure french, hsdb 438
trifluorostibane

Specifications

Antimony trifluoride
7783-56-4
F3Sb
MFCD00011218
15, 697
Solubility in water: 3850g/L (20°C). Other solubilities: decomposes in water,insoluble in nh3
F[Sb](F)F
178.75
178.75
98%
Solubility in water: 3850g/L (20°C). Other solubilities: decomposes in water, insoluble in nh3
376.0°C
292.0°C
98%
97.5% min. (Iodimetry)
SbF3
05,526; 06,553
antimony trifluoride, trifluorostibine, antimony fluoride, antimony iii fluoride, stibine, trifluoro, trifluoroantimony, antimoine fluorure, antimonous fluoride, antimoine fluorure french, hsdb 438
GUNJVIDCYZYFGV-UHFFFAOYSA-K
trifluorostibane
24554
Crystalline Powder
Plastic bottle
376 °C
White to Gray
100g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Antimony trifluoride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents